Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022413.4 | NP_071858.3 | 15 | 2207 | |
GenBank mRNA | AB037373.1 | - | 15 | 2114 | |
AJ542487.1 | - | 15 | 1912 | ||
AK077234.1 | - | 15 | 2137 | ||
AK168760.1 | - | 15 | 2205 | ||
BC016101.1 | - | 15 | 2145 | 4911355 |