Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172481.1 | NP_001165952.1 | 4 | 748 | |
NM_025711.3 | NP_079987.2 | 4 | 765 | ||
GenBank mRNA | AF316825.1 | - | 3 | 485 | |
AK014504.1 | - | 4 | 721 | ||
AK085920.1 | - | 4 | 720 | ||
BC034888.1 | - | 4 | 667 | 1365428 | |
BC145904.1 | - | 4 | 663 | 40131207 | |
BC145930.1 | - | 4 | 646 | 40131233 |