Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023343.2 | NP_075832.1 | 9 | 866 | |
GenBank mRNA | AF117831.1 | - | 4 | 320 | |
AK003007.1 | - | 8 | 1839 | ||
AK005408.1 | - | 9 | 889 | ||
AK019158.1 | - | 3 | 250 | ||
BC011427.1 | - | 9 | 836 | ||
BC026953.1 | - | 9 | 866 | 5133980 | |
BC027439.1 | - | 9 | 755 | 4164301 |