Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000242.2 | NP_000233.1 | 4 | 661 | |
XM_006717861.3 | XP_006717924.1 | 5 | 729 | ||
XM_011539816.2 | XP_011538118.1 | 5 | 713 | ||
GenBank mRNA | AF360991.1 | - | 4 | 612 | |
AY826184.1 | - | 4 | 596 | ||
BC069338.1 | - | 4 | 646 | 7262231 | |
BC096179.3 | - | 4 | 627 | 40003735 | |
BC096180.3 | - | 4 | 627 | 40003741 | |
BC096181.3 | - | 4 | 627 | 40003742 | |
BC096182.3 | - | 4 | 627 | 40003743 | |
X15422.1 | - | 4 | 661 |