Gene Transcripts

Gene Symbol
Fam173b
Entrez Gene ID
68073
Gene Name
family with sequence similarity 173, member B
Gene Aliases
A930016P21Rik, AA987072
Chromosome Location
Chr. 15: 31601941 - 31617537 on Build GRCm38
UniGene ID
Mm.119185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026546.3 NP_080822.1 3, 4 457
GenBank mRNA AK020868.1 - 3, 4 457
AK160923.1 - 584
BC030179.1 - 2, 3 448 5039149

View Full Product Details