Gene Transcripts
- Gene Symbol
- Snhg17
- Entrez Gene ID
- 68108
- Gene Name
- small nucleolar RNA host gene 17
- Gene Aliases
- 9430008C03Rik
- Chromosome Location
- Chr. 2: 158353700 - 158361522 on Build GRCm38
- UniGene ID
- Mm.119198
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007971.1 |
- |
4, 5 |
541 |
|
AK046044.1 |
- |
4, 5 |
747 |
|
AK079109.1 |
- |
3, 4 |
1673 |
|
View Full Product Details