Gene Transcripts

Gene Symbol
Cyp4f16
Entrez Gene ID
70101
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 16
Gene Aliases
2310021J05Rik, AI746515
Chromosome Location
Chr. 17: 32536545 - 32551798 on Build GRCm38
UniGene ID
Mm.30504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024442.1 NP_077762.1 3 254
GenBank mRNA AF233646.1 - 3 254
AK078557.1 - 3 315
AK146232.1 - 3 327
BC026539.1 - 3 286 4982322

View Full Product Details