Gene Transcripts

Gene Symbol
Ist1
Entrez Gene ID
71955
Gene Name
increased sodium tolerance 1 homolog (yeast)
Gene Aliases
2400003C14Rik, AW536298, mKIAA0174
Chromosome Location
Chr. 8: 109671321 - 109693294 on Build GRCm38
UniGene ID
Mm.290036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028018.2 NP_082294.1 5, 6 560
GenBank mRNA AK010262.1 - 5, 6 530
AK050468.1 - 5, 6 527
AK088192.1 - 5, 6 515
AK122215.1 - 3, 4 334
AK152830.1 - 5, 6 527
AK161899.1 - 5, 6 516
AK207302.1 - 3, 4 254
BC017605.1 - 5, 6 526 4507347
BC039052.1 - 5, 6 503 2631157

View Full Product Details