Gene Transcripts
- Gene Symbol
- DHPS
- Entrez Gene ID
- 1725
- Gene Name
- deoxyhypusine synthase
- Gene Aliases
- DHS, DS, MIG13
- Chromosome Location
- Chr. 19: 12672164 - 12681887 on Build GRCh38
- UniGene ID
- Hs.79064
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206974.1 |
NP_001193903.1
|
3, 4 |
465 |
|
NM_001930.3 |
NP_001921.1
|
3, 4 |
614 |
|
NM_013406.2 |
NP_037538.1
|
3, 4 |
614 |
|
NR_038192.1 |
|
3, 4 |
614 |
|
XM_011527770.1 |
XP_011526072.1
|
3, 4 |
614 |
|
XM_011527771.2 |
XP_011526073.1
|
3, 4 |
625 |
|
GenBank mRNA |
AK291553.1 |
- |
3, 4 |
595 |
|
AK304780.1 |
- |
3, 4 |
560 |
|
AL520040.3 |
- |
3, 4 |
390 |
|
AY336090.1 |
- |
3, 4 |
503 |
|
BC000333.2 |
- |
3, 4 |
561 |
2822554 |
BC014016.2 |
- |
3, 4 |
576 |
4129051 |
BU844677.1 |
- |
3, 4 |
593 |
|
L39068.1 |
- |
3, 4 |
530 |
|
U26266.1 |
- |
3, 4 |
495 |
|
U32178.1 |
- |
3, 4 |
495 |
|
U40579.1 |
- |
3, 4 |
576 |
|
U79262.1 |
- |
3, 4 |
588 |
|
View Full Product Details