Gene Transcripts
- Gene Symbol
- Fance
- Entrez Gene ID
- 72775
- Gene Name
- Fanconi anemia, complementation group E
- Gene Aliases
- 2810451D06Rik, AI415634, AW209126
- Chromosome Location
- Chr. 17: 28313530 - 28326574 on Build GRCm38
- UniGene ID
- Mm.201859
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163819.1 |
NP_001157291.1
|
8, 9 |
1467 |
|
GenBank mRNA |
AK135017.1 |
- |
9, 10 |
1342 |
|
AK172283.1 |
- |
7, 8 |
1594 |
|
AK182233.1 |
- |
1, 2 |
36 |
|
AK184214.1 |
- |
1, 2 |
35 |
|
AK186303.1 |
- |
1, 2 |
35 |
|
AK188566.1 |
- |
1, 2 |
35 |
|
AK207586.1 |
- |
1, 2 |
35 |
|
AK219855.1 |
- |
1, 2 |
35 |
|
BC039987.1 |
- |
8, 9 |
1401 |
|
View Full Product Details