Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027261.3 | NP_081537.1 | 3 | 454 | |
NM_029248.2 | NP_083524.2 | 3 | 454 | ||
GenBank mRNA | AK012656.1 | - | 1 | 304 | |
AK012662.1 | - | 3 | 406 | ||
AK014920.1 | - | 3 | 406 | ||
AK016109.1 | - | 3 | 408 | ||
AK050165.1 | - | 3 | 410 | ||
AK161656.1 | - | 3 | 409 | ||
AK166329.1 | - | 3 | 409 | ||
AK166633.1 | - | 3 | 409 | ||
BC027813.1 | - | 1 | 242 | ||
BC056964.1 | - | 2 | 392 | ||
BC110660.1 | - | 2 | 396 | 3158156 | |
BC117068.1 | - | 2 | 386 | 40126387 | |
BC117070.1 | - | 2 | 386 | 40126389 |