Gene Transcripts

Gene Symbol
Nme5
Entrez Gene ID
75533
Gene Name
NME/NM23 family member 5
Gene Aliases
1700019D05Rik, Nm23-M5
Chromosome Location
Chr. 18: 34562636 - 34579152 on Build GRCm38
UniGene ID
Mm.275430
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080637.3 NP_542368.2 4, 5 553
GenBank mRNA AF343565.1 - 3, 4 442
AK131722.1 - 4, 5 554

View Full Product Details