Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178898.4 | NP_849229.2 | 5 | 1827 | |
GenBank mRNA | AK040541.1 | - | 5 | 1893 | |
AK045736.1 | - | 5 | 1816 | ||
AK133549.1 | - | 5 | 1828 | ||
AK162106.1 | - | 5 | 1756 | ||
BC090657.1 | - | 5 | 1769 | ||
BC096536.1 | - | 4 | 1537 | ||
BC119280.1 | - | 5 | 1705 | 8734029 | |
BC120581.1 | - | 5 | 1706 | 40129504 | |
BC144752.1 | - | 5 | 1706 | 9053300 |