Gene Transcripts

Gene Symbol
G6b
Entrez Gene ID
106722
Gene Name
immunoreceptor tyrosine-based inhibitory motif (ITIM) containing platelet receptor
Gene Aliases
AU023871, C6orf25, Ng31
Chromosome Location
Chr. 17: 35062693 - 35066785 on Build GRCm38
UniGene ID
Mm.390956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033221.3 NP_001028393.1 2 352
NM_001191012.1 NP_001177941.1 2 352
GenBank mRNA AK137516.1 - 2 353
GU269270.1 - 2 338

View Full Product Details