Gene Transcripts

Gene Symbol
Afap1l1
Entrez Gene ID
106877
Gene Name
actin filament associated protein 1-like 1
Gene Aliases
AI173486
Chromosome Location
Chr. 18: 61730262 - 61786662 on Build GRCm38
UniGene ID
Mm.26632
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178928.4 NP_849259.2 16 1938
GenBank mRNA AK035227.1 - 16 1938
AK053714.1 - 15 2173
BC094505.1 - 13 1376
BC138923.1 - 16 1899 8861945

View Full Product Details