Gene Transcripts

Gene Symbol
Smim3
Entrez Gene ID
106878
Gene Name
small integral membrane protein 3
Gene Aliases
2010002N04Rik, AI195350, AW561905, Nid67, cI-41
Chromosome Location
Chr. 18: 60474191 - 60518947 on Build GRCm38
UniGene ID
Mm.273197
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134133.2 NP_598894.1 2 737
GenBank mRNA AF313412.1 - 2 279
AK143383.1 - 2 741
AK155873.1 - 2 383
AK168283.1 - 2 561
AK171810.1 - 2 395
BC012898.1 - 2 245 4013357

View Full Product Details