Gene Transcripts

Gene Symbol
Amy2a5
Entrez Gene ID
109959
Gene Name
amylase 2a5
Gene Aliases
1810008N23Rik, Amy-2, Amy2, mAmy2-2
Chromosome Location
Chr. 3: 113249178 - 113258699 on Build GRCm38
UniGene ID
Mm.275426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042711.2 NP_001036176.1 260
GenBank mRNA AK078968.1 - 258
AK133526.1 - 261
BC094924.1 - 248 6433956
BC100579.1 - 255 6433256
V00718.1 - 261
X02576.1 - 259
X02577.1 - 259
X02578.1 - 259
X02579.1 - 259

Gene Symbol
Amy2a2
Entrez Gene ID
100043688
Gene Name
amylase 2a2
Gene Aliases
ENSMUSG00000074265, OTTMUSG00000022431, mAmy2-5
Chromosome Location
Chr. 3: 113345049 - 113356658 on Build GRCm38
UniGene ID
Mm.275426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160152.1 NP_001153624.1 2 398

Gene Symbol
Amy2a3
Entrez Gene ID
100043686
Gene Name
amylase 2a3
Gene Aliases
OTTMUSG00000022430, mAmy2-4
Chromosome Location
Chr. 3: 113312449 - 113324052 on Build GRCm38
UniGene ID
Mm.275426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160151.1 NP_001153623.1 398

Gene Symbol
Amy2a4
Entrez Gene ID
100043684
Gene Name
amylase 2a4
Gene Aliases
ENSMUSG00000074267, OTTMUSG00000022433, mAmy2-3
Chromosome Location
Chr. 3: 113279837 - 113291449 on Build GRCm38
UniGene ID
Mm.275426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160150.1 NP_001153622.1 398

View Full Product Details