Gene Transcripts

Gene Symbol
APCS
Entrez Gene ID
325
Gene Name
amyloid P component, serum
Gene Aliases
HEL-S-92n, PTX2, SAP
Chromosome Location
Chr. 1: 159587826 - 159588871 on Build GRCh38
UniGene ID
Hs.507080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001639.3 NP_001630.1 2 182
GenBank mRNA BC007039.1 - 2 182
BC007058.1 - 2 182 3934872
BT006750.1 - 2 86
CR450313.1 - 2 86
FJ460512.1 - 2 182
M10944.1 - 2 86
X04608.1 - 2 182

View Full Product Details