Gene Transcripts
- Gene Symbol
- DNAJA1
- Entrez Gene ID
- 3301
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A1
- Gene Aliases
- DJ-2, DjA1, HDJ2, HSDJ, HSJ-2, HSJ2, HSPF4, NEDD7, hDJ-2
- Chromosome Location
- Chr. 9: 33025203 - 33039907 on Build GRCh38
- UniGene ID
- Hs.445203
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001539.3 |
NP_001530.1
|
2, 3 |
323 |
|
GenBank mRNA |
AK289623.1 |
- |
2, 3 |
251 |
|
AY186741.1 |
- |
2, 3 |
249 |
|
BC008182.1 |
- |
2, 3 |
236 |
2900892 |
BI462555.1 |
- |
2, 3 |
318 |
|
BM469197.1 |
- |
2, 3 |
224 |
|
BT007292.1 |
- |
1, 2 |
132 |
|
D13388.1 |
- |
2, 3 |
249 |
|
DA003255.1 |
- |
2, 3 |
325 |
|
L08069.1 |
- |
2, 3 |
214 |
|
View Full Product Details