Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001663.3 | NP_001654.1 | 2 | 1185 | |
GenBank mRNA | AF047432.1 | - | 1 | 631 | |
BC002952.1 | - | 2 | 1088 | ||
BC008918.2 | - | 2 | 1100 | 3626100 | |
M57763.1 | - | 2 | 1081 |