Gene Transcripts

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
Chromosome Location
Chr. 9: 17906574 - 18910950 on Build GRCh38
UniGene ID
Hs.741483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040272.5 NP_001035362.3 4 571
NM_052866.4 NP_443098.3 4 571
XM_011518063.2 XP_011516365.1 6 709
XM_011518064.2 XP_011516366.1 5 824
XM_017015310.1 XP_016870799.1 5 7346
XM_017015311.1 XP_016870800.1 6 709
XM_017015312.1 XP_016870801.1 5 825
XM_017015313.1 XP_016870802.1 5 472
XM_017015314.1 XP_016870803.1 6 709
GenBank mRNA AF176313.1 - 4 469
AK290667.1 - 4 557
AK291956.1 - 4 511
AY358327.1 - 4 551
BC030262.1 - 4 483 5201199
CX758410.1 - 4 470

View Full Product Details