Gene Transcripts

Gene Symbol
TRPA1
Entrez Gene ID
8989
Gene Name
transient receptor potential cation channel subfamily A member 1
Gene Aliases
ANKTM1, FEPS
Chromosome Location
Chr. 8: 72021251 - 72094885 on Build GRCh38
UniGene ID
Hs.716816
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007332.2 NP_015628.2 17 2211
XM_011517624.2 XP_011515926.1 18 7027
XM_011517625.2 XP_011515927.1 19 7186
XM_017013946.1 XP_016869435.1 18 3489
GenBank mRNA AK307942.1 - 17 2226
Y10601.1 - 17 2210

View Full Product Details