Gene Transcripts

Gene Symbol
PSEN1
Entrez Gene ID
5663
Gene Name
presenilin 1
Gene Aliases
AD3, FAD, PS-1, PS1, S182
Chromosome Location
Chr. 14: 73136435 - 73223691 on Build GRCh38
UniGene ID
Hs.3260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000021.3 NP_000012.1 8 1088
NM_007318.2 NP_015557.2 8 1076
XM_005267864.2 XP_005267921.1 8 1082
XM_005267866.1 XP_005267923.1 8 1071
XM_011536971.2 XP_011535273.1 8 980
XM_011536972.2 XP_011535274.1 8 1344
XM_011536973.1 XP_011535275.1 8 1332
XM_011536974.1 XP_011535276.1 8 973
GenBank mRNA AF416717.1 - 6 804
AK122722.1 - 6 2357
AK223173.1 - 6 818
AK312531.1 - 8 1072
BC011729.2 - 8 1057 4423098
KU178285.1 - 6 792
KU178286.1 - 6 804
L42110.1 - 8 1052
L76517.1 - 8 1345
U40379.1 - 6 792
U40380.1 - 6 792

View Full Product Details