Gene Transcripts

Gene Symbol
USP37
Entrez Gene ID
57695
Gene Name
ubiquitin specific peptidase 37
Gene Aliases
Chromosome Location
Chr. 2: 218450251 - 218568393 on Build GRCh38
UniGene ID
Hs.166068
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020935.2 NP_065986.2 26 7407
XM_005246720.2 XP_005246777.1 26 7479
XM_005246721.3 XP_005246778.1 26 7371
XM_005246722.3 XP_005246779.1 26 7411
XM_005246724.1 XP_005246781.1 25 7341
XM_011511538.2 XP_011509840.1 25 7352
XM_017004577.1 XP_016860066.1 25 7267
GenBank mRNA BC037389.1 - 1 3081
BX538024.1 - 26 7379

View Full Product Details