Gene Transcripts

Gene Symbol
AFG3L1P
Entrez Gene ID
172
Gene Name
AFG3 like matrix AAA peptidase subunit 1, pseudogene
Gene Aliases
AFG3, AFG3L1
Chromosome Location
Chr. 16: 89972580 - 90000787 on Build GRCh38
UniGene ID
Hs.534773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003226.1 6 723
NR_003227.1 6 650
NR_003228.1 6 689
GenBank mRNA AF329691.1 - 5 474
AF329693.1 - 5 474
AF329694.1 - 4 405
AK056488.1 - 6 676
AK091293.1 - 6 686
AK097044.1 - 6 637
AK126911.1 - 5 614
AK128804.1 - 3 262
BC021245.2 - 4 296

View Full Product Details