Gene Transcripts
- Gene Symbol
- MMEL1
- Entrez Gene ID
- 79258
- Gene Name
- membrane metallo-endopeptidase-like 1
- Gene Aliases
- MMEL2, NEP2, NEPII, NL1, NL2, SEP
- Chromosome Location
- Chr. 1: 2590639 - 2633042 on Build GRCh38
- UniGene ID
- Hs.591453
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033467.3 |
NP_258428.2
|
5, 6 |
676 |
|
XM_017002310.1 |
XP_016857799.1
|
6, 7 |
942 |
|
XM_017002311.1 |
XP_016857800.1
|
5, 6 |
676 |
|
XM_017002312.1 |
XP_016857801.1
|
4, 5 |
594 |
|
XM_017002313.1 |
XP_016857802.1
|
5, 6 |
676 |
|
XM_017002314.1 |
XP_016857803.1
|
5, 6 |
676 |
|
XM_017002315.1 |
XP_016857804.1
|
5, 6 |
552 |
|
GenBank mRNA |
AF336981.1 |
- |
4, 5 |
499 |
|
AK093058.1 |
- |
5, 6 |
624 |
|
BC101027.2 |
- |
4, 5 |
472 |
40007698 |
BC101028.1 |
- |
4, 5 |
472 |
40007699 |
BC101029.2 |
- |
4, 5 |
472 |
40007702 |
BC101030.1 |
- |
4, 5 |
472 |
40007706 |
View Full Product Details