Gene Transcripts
- Gene Symbol
- PSMB6
- Entrez Gene ID
- 5694
- Gene Name
- proteasome subunit beta 6
- Gene Aliases
- DELTA, LMPY, Y
- Chromosome Location
- Chr. 17: 4796144 - 4798503 on Build GRCh38
- UniGene ID
- Hs.77060
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270481.1 |
NP_001257410.1
|
3, 4 |
361 |
|
NM_002798.2 |
NP_002789.1
|
3, 4 |
361 |
|
GenBank mRNA |
AK311768.1 |
- |
3, 4 |
324 |
|
BC000835.2 |
- |
3, 4 |
328 |
3456009 |
BF671774.1 |
- |
3, 4 |
362 |
|
BM553399.1 |
- |
3, 4 |
308 |
|
BQ691763.1 |
- |
3, 4 |
343 |
|
BT009809.1 |
- |
3, 4 |
310 |
|
BU848746.1 |
- |
3, 4 |
293 |
|
CR457065.1 |
- |
3, 4 |
310 |
|
D29012.1 |
- |
3, 4 |
319 |
|
X61971.1 |
- |
1, 2 |
133 |
|
View Full Product Details