Gene Transcripts

Gene Symbol
DEFA4
Entrez Gene ID
1669
Gene Name
defensin alpha 4
Gene Aliases
DEF4, HNP-4, HP-4, HP4
Chromosome Location
Chr. 8: 6935820 - 6938338 on Build GRCh38
UniGene ID
Hs.591391
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001925.2 NP_001916.1 2 253
GenBank mRNA BC093959.1 - 2 177
BC106747.2 - 1 148 40021796
BC112091.1 - 2 177 8327559
DN998607.1 - 2 202
X65977.1 - 2 179

View Full Product Details