Gene Transcripts
- Gene Symbol
- PMM2
- Entrez Gene ID
- 5373
- Gene Name
- phosphomannomutase 2
- Gene Aliases
- CDG1, CDG1a, CDGS, PMI, PMI1, PMM 2
- Chromosome Location
- Chr. 16: 8797813 - 8849337 on Build GRCh38
- UniGene ID
- Hs.625732
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000303.2 |
NP_000294.1
|
7, 8 |
718 |
|
XM_005255374.4 |
XP_005255431.1
|
6, 7 |
641 |
|
GenBank mRNA |
AB209659.1 |
- |
3, 4 |
3816 |
|
AK291537.1 |
- |
7, 8 |
689 |
|
AK296630.1 |
- |
5, 6 |
495 |
|
AK299979.1 |
- |
6, 7 |
612 |
|
AK300785.1 |
- |
6, 7 |
594 |
|
AK304320.1 |
- |
4, 5 |
412 |
|
AK311035.1 |
- |
5, 6 |
512 |
|
AK312532.1 |
- |
7, 8 |
689 |
|
BC008310.1 |
- |
7, 8 |
660 |
3611382 |
KU178265.1 |
- |
7, 8 |
648 |
|
KU178266.1 |
- |
5, 6 |
479 |
|
U85773.1 |
- |
7, 8 |
696 |
|
View Full Product Details