Gene Transcripts

Gene Symbol
TRIM21
Entrez Gene ID
6737
Gene Name
tripartite motif containing 21
Gene Aliases
RNF81, RO52, Ro/SSA, SSA, SSA1
Chromosome Location
Chr. 11: 4384897 - 4393696 on Build GRCh38
UniGene ID
Hs.532357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003141.3 NP_003132.2 6, 7 969
GenBank mRNA AK225564.1 - 4, 5 682
AY742713.1 - 5, 6 856
BC010861.1 - 6, 7 940 3900540
M34551.1 - 5, 6 894
M62800.1 - 6, 7 941

View Full Product Details