Gene Transcripts

Gene Symbol
VEZF1
Entrez Gene ID
7716
Gene Name
vascular endothelial zinc finger 1
Gene Aliases
DB1, ZNF161
Chromosome Location
Chr. 17: 57971547 - 57988254 on Build GRCh38
UniGene ID
Hs.463569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007146.2 NP_009077.2 6 1961
XM_005257643.1 XP_005257700.1 6 1958
XM_005257644.4 XP_005257701.1 7 2114
XM_017025017.1 XP_016880506.1 7 2165
XM_017025018.1 XP_016880507.1 7 2017
GenBank mRNA BX648862.1 - 6 1254
D28118.1 - 6 1844

View Full Product Details