Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318192.1 | NP_001305121.1 | 12 | 1944 | |
NM_012450.3 | NP_036582.2 | 12 | 1941 | ||
XM_011516024.2 | XP_011514326.1 | 12 | 1724 | ||
XM_017011962.1 | XP_016867451.1 | 10 | 1312 | ||
GenBank mRNA | AB208809.1 | - | 9 | 6219 | |
AF169301.1 | - | 12 | 1914 | ||
AK313072.1 | - | 12 | 1534 | ||
BC030689.2 | - | 12 | 1923 | 4821193 |