Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr. 7: 128954180 - 129055173 on Build GRCh38
UniGene ID
Hs.193613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191028.2 NP_001177957.2 6 1113
NM_012470.3 NP_036602.1 6 1113
NR_034053.2 7 1274
XM_011515989.2 XP_011514291.1 6 715
GenBank mRNA AF145029.1 - 6 823
AJ133769.1 - 6 710
AK225999.1 - 6 1084
AK312580.1 - 6 840
BC068530.1 - 6 841

View Full Product Details