Gene Transcripts
- Gene Symbol
- PILRB
- Entrez Gene ID
- 29990
- Gene Name
- paired immunoglobin-like type 2 receptor beta
- Gene Aliases
- FDFACT1, FDFACT2
- Chromosome Location
- Chr. 7: 100358003 - 100367831 on Build GRCh38
- UniGene ID
- Hs.632314
- Species
- Homo sapiens
- Gene Symbol
- STAG3L5P-PVRIG2P-PILRB
- Entrez Gene ID
- 101752399
- Gene Name
- STAG3L5P-PVRIG2P-PILRB readthrough
- Gene Aliases
- LOC101735302-LOC101752334-PILRB
- Chromosome Location
- Chr. 7: 100336065 - 100367831 on Build GRCh38
- UniGene ID
- Hs.521075
- Species
- Homo sapiens
View Full Product Details