Gene Transcripts

Gene Symbol
LEPROTL1
Entrez Gene ID
23484
Gene Name
leptin receptor overlapping transcript-like 1
Gene Aliases
HSPC112, Vps55, my047
Chromosome Location
Chr. 8: 30095406 - 30137706 on Build GRCh38
UniGene ID
Hs.146585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015344.2 NP_056159.2 4 2244
GenBank mRNA AF063605.1 - 4 2221
AK075363.1 - 4 2189
AY358674.1 - 4 2216

View Full Product Details