Gene Transcripts

Gene Symbol
LMO3
Entrez Gene ID
55885
Gene Name
LIM domain only 3
Gene Aliases
RBTN3, RBTNL2, RHOM3, Rhom-3
Chromosome Location
Chr. 12: 16548372 - 16610233 on Build GRCh38
UniGene ID
Hs.504908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001395.2 NP_001001395.1 4 724
NM_001243609.1 NP_001230538.1 5 791
NM_001243610.1 NP_001230539.1 5 563
NM_001243611.1 NP_001230540.1 4 960
NM_001243612.1 NP_001230541.1 4 648
NM_001243613.1 NP_001230542.1 6 728
NM_018640.4 NP_061110.2 4 982
NR_045012.1 6 916
NR_045013.1 5 732
NR_045014.1 4 665
XM_006719110.3 XP_006719173.1 5 799
XM_006719111.2 XP_006719174.1 4 842
XM_011520762.1 XP_011519064.1 5 690
XM_011520763.1 XP_011519065.1 5 597
XM_017019643.1 XP_016875132.1 5 1143
GenBank mRNA AB044745.1 - 4 728
AB044746.1 - 6 629
AF258348.1 - 5 520
AK095595.1 - 4 600
AK294474.1 - 4 648
AK294909.1 - 6 728
AK308594.1 - 7 1076
AK308826.1 - 7 792
AK308896.1 - 6 728
AK316021.1 - 4 584
AL050152.1 - 1 19
BC017777.1 - 4 478
BC026311.1 - 4 878 4799836
BC050085.1 - 5 564 5274811
BI553855.1 - 4 621

View Full Product Details