Gene Transcripts

Gene Symbol
LMO3
Entrez Gene ID
55885
Gene Name
LIM domain only 3
Gene Aliases
RBTN3, RBTNL2, RHOM3, Rhom-3
Chromosome Location
Chr. 12: 16548372 - 16610233 on Build GRCh38
UniGene ID
Hs.504908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001395.2 NP_001001395.1 4 851
NM_001243609.1 NP_001230538.1 5 918
NM_001243610.1 NP_001230539.1 5 690
NM_001243611.1 NP_001230540.1 4 1087
NM_001243612.1 NP_001230541.1 4 775
NM_001243613.1 NP_001230542.1 6 855
NM_018640.4 NP_061110.2 4 1109
NR_045012.1 6 1043
NR_045013.1 5 859
NR_045014.1 4 792
XM_006719110.3 XP_006719173.1 5 926
XM_006719111.2 XP_006719174.1 4 969
XM_011520762.1 XP_011519064.1 5 817
XM_011520763.1 XP_011519065.1 5 724
XM_017019643.1 XP_016875132.1 5 1270
GenBank mRNA AB044745.1 - 4 855
AF258348.1 - 5 647
AK095595.1 - 4 727
AK294474.1 - 4 775
AK294909.1 - 6 855
AK308594.1 - 7 1203
AK308896.1 - 6 855
AK316021.1 - 4 711
AL050152.1 - 1 146
BC017777.1 - 4 604
BC026311.1 - 4 1005 4799836
BC050085.1 - 5 691 5274811

View Full Product Details