Gene Transcripts

Gene Symbol
TREM1
Entrez Gene ID
54210
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
CD354, TREM-1
Chromosome Location
Chr. 6: 41267385 - 41286745 on Build GRCh38
UniGene ID
Hs.283022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242589.2 NP_001229518.1 2 349
NM_001242590.2 NP_001229519.1 2 349
NM_018643.4 NP_061113.1 2 349
NR_136332.1 2 349
XM_006715117.3 XP_006715180.1 2 286
XM_011514696.1 XP_011512998.1 2 289
XM_011514697.1 XP_011512999.1 2 286
XM_017010956.1 XP_016866445.1 2 289
XM_017010957.1 XP_016866446.1 2 286
GenBank mRNA AF196329.1 - 2 306
AF287008.1 - 2 323
AK223264.1 - 2 286
AK292162.1 - 2 286
AK301519.1 - 2 286
AK313104.1 - 2 301
AY074783.1 - 2 276
BC017773.1 - 2 304 4692680
BI910026.1 - 2 269
DC428714.1 - 2 349

View Full Product Details