Gene Transcripts

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
Chromosome Location
Chr. 8: 103398638 - 103415335 on Build GRCh38
UniGene ID
Hs.532265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030780.4 NP_110407.2 7 1246
NR_102337.1 6 1160
NR_102338.1 8 1355
XM_017013877.1 XP_016869366.1 7 1149
GenBank mRNA AF283645.1 - 7 976
AK027531.1 - 7 1015
AK027787.1 - 7 980
AK125492.1 - 8 964
AK300748.1 - 6 894
BC021893.1 - 7 954 4524907

View Full Product Details