Gene Transcripts
- Gene Symbol
- RTN4IP1
- Entrez Gene ID
- 84816
- Gene Name
- reticulon 4 interacting protein 1
- Gene Aliases
- NIMP, OPA10
- Chromosome Location
- Chr. 6: 106559237 - 106630921 on Build GRCh38
- UniGene ID
- Hs.155839
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318746.1 |
NP_001305675.1
|
4, 5 |
512 |
|
NM_032730.5 |
NP_116119.2
|
4, 5 |
1098 |
|
XM_011536192.2 |
XP_011534494.1
|
5, 6 |
1186 |
|
XM_017011375.1 |
XP_016866864.1
|
4, 5 |
1098 |
|
XM_017011376.1 |
XP_016866865.1
|
4, 5 |
1098 |
|
GenBank mRNA |
AF336861.1 |
- |
4, 5 |
672 |
|
AK095207.1 |
- |
4, 5 |
512 |
|
AK313962.1 |
- |
4, 5 |
1016 |
|
AY063761.1 |
- |
4, 5 |
621 |
|
BC006399.2 |
- |
4, 5 |
893 |
4309263 |
View Full Product Details