Gene Transcripts

Gene Symbol
LRWD1
Entrez Gene ID
222229
Gene Name
leucine rich repeats and WD repeat domain containing 1
Gene Aliases
CENP-33, ORCA
Chromosome Location
Chr. 7: 102464883 - 102473168 on Build GRCh38
UniGene ID
Hs.274135
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317721.1 NP_001304650.1 15 2192
NM_152892.2 NP_690852.1 15 2174
XM_017011845.1 XP_016867334.1 15 2129
GenBank mRNA AB209379.1 - 7 5406
AK290967.1 - 15 2073
AL133057.1 - 15 2124
BC009436.1 - 14 1865
BC018769.1 - 14 1704
BC030547.1 - 15 2114 5170588

View Full Product Details