Gene Transcripts

Gene Symbol
LRRC57
Entrez Gene ID
255252
Gene Name
leucine rich repeat containing 57
Gene Aliases
Chromosome Location
Chr. 15: 42528332 - 42548804 on Build GRCh38
UniGene ID
Hs.234681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153260.2 NP_694992.2 6 1056
GenBank mRNA AK094131.1 - 6 1050
AK094891.1 - 5 1290
BX538324.1 - 5 1202
CN350663.1 - 3 506
CR749223.1 - 6 1025

View Full Product Details