Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040202.1 | NP_001035292.1 | 6 | 1300 | |
GenBank mRNA | AK055774.1 | - | 6 | 1088 | |
AK126273.1 | - | 3 | 505 | ||
AX775925.1 | - | 6 | 1294 | ||
AY424281.1 | - | 6 | 1227 | ||
BC047510.1 | - | 7 | 1556 | 5297045 | |
BX648346.1 | - | 5 | 931 |