Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320289.1 | NP_001307218.1 | 9 | 1340 | |
NM_001320290.1 | NP_001307219.1 | 9 | 1077 | ||
NM_178033.1 | NP_828847.1 | 9 | 1374 | ||
XM_017000973.1 | XP_016856462.1 | 9 | 1405 | ||
GenBank mRNA | AK091806.1 | - | 9 | 1078 | |
AK098065.1 | - | 9 | 1367 | ||
AK131355.1 | - | 9 | 1340 | ||
AM040940.1 | - | 9 | 1211 | ||
AY358537.1 | - | 9 | 1357 | ||
BC028102.1 | - | 9 | 1374 | 5243005 |