Gene Transcripts

Gene Symbol
GPI
Entrez Gene ID
2821
Gene Name
glucose-6-phosphate isomerase
Gene Aliases
AMF, GNPI, NLK, PGI, PHI, SA-36, SA36
Chromosome Location
Chr. 19: 34359592 - 34402413 on Build GRCh38
UniGene ID
Hs.466471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000175.3 NP_000166.2 2 285
NM_001184722.1 NP_001171651.1 3 432
NM_001289789.1 NP_001276718.1 3 432
NM_001289790.1 NP_001276719.1 2 285
XM_005258764.2 XP_005258821.1 3 251
XM_006723148.2 XP_006723211.1 3 202
XM_011526754.2 XP_011525056.1 4 667
GenBank mRNA AB209575.1 - 1 1134
AK129884.1 - 3 216
AK293446.1 - 2 161
AK294396.1 - 3 380
AK301103.1 - 2 170
BC004982.1 - 2 153 2906270
DC299841.1 - 2 285
DC342043.1 - 3 432
K03515.1 - 2 160
X16539.1 - 2 148

View Full Product Details