Gene Transcripts
- Gene Symbol
- TPST1
- Entrez Gene ID
- 8460
- Gene Name
- tyrosylprotein sulfotransferase 1
- Gene Aliases
- TANGO13A
- Chromosome Location
- Chr. 7: 66205272 - 66360451 on Build GRCh38
- UniGene ID
- Hs.421194
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC013188.1 |
- |
|
2010 |
|
View Full Product Details