Gene Transcripts
- Gene Symbol
- AKR7L
- Entrez Gene ID
- 246181
- Gene Name
- aldo-keto reductase family 7 like (gene/pseudogene)
- Gene Aliases
- AFAR3, AFB1-AR3, AKR7A4
- Chromosome Location
- Chr. 1: 19265982 - 19274074 on Build GRCh38
- UniGene ID
- Hs.662342
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040288.1 |
|
7 |
2079 |
|
NR_040289.1 |
|
5 |
1882 |
|
GenBank mRNA |
BC035351.1 |
- |
5 |
1748 |
|
View Full Product Details