Gene Transcripts

Gene Symbol
IDH2
Entrez Gene ID
3418
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Gene Aliases
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
Chromosome Location
Chr. 15: 90083978 - 90102554 on Build GRCh38
UniGene ID
Hs.596461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289910.1 NP_001276839.1 3 173
NM_002168.3 NP_002159.2 3 406
GenBank mRNA AK222916.1 - 3 320
AK294148.1 - 3 173
AK312627.1 - 3 370
AK316388.1 - 3 247
BC009244.2 - 3 308 2959540
BC071828.1 - 3 307 6090792
HY010337.1 - 3 406
X69433.1 - 3 328

View Full Product Details