Gene Transcripts

Gene Symbol
LY6H
Entrez Gene ID
4062
Gene Name
lymphocyte antigen 6 complex, locus H
Gene Aliases
NMLY6
Chromosome Location
Chr. 8: 143157914 - 143160636 on Build GRCh38
UniGene ID
Hs.159590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130478.1 NP_001123950.1 5 421
NM_001135655.1 NP_001129127.1 4 401
NM_002347.4 NP_002338.3 4 365
XM_017013413.1 XP_016868902.1 3 379
XM_017013414.1 XP_016868903.1 4 426
GenBank mRNA AB012293.1 - 4 345
AK314139.1 - 5 421
BC028894.1 - 4 391 4939118
BC030192.1 - 4 325 5199071
BI914261.1 - 3 206
BP347868.1 - 4 329
CR457034.1 - 3 199
DB471677.1 - 4 365
DB502453.1 - 4 414

View Full Product Details