Gene Transcripts

Gene Symbol
TRPV3
Entrez Gene ID
162514
Gene Name
transient receptor potential cation channel subfamily V member 3
Gene Aliases
FNEPPK2, OLMS, VRL3
Chromosome Location
Chr. 17: 3510502 - 3557995 on Build GRCh38
UniGene ID
Hs.446255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258205.1 NP_001245134.1 7, 8 1098
NM_145068.3 NP_659505.1 7, 8 1098
GenBank mRNA AF514998.1 - 7, 8 840
AJ487035.2 - 7, 8 1098
AK127726.1 - 1, 2 50
AY118267.1 - 7, 8 908
AY118268.1 - 7, 8 908
BC104866.1 - 7, 8 870 8143869
BC104868.1 - 7, 8 870 8143871
BC143294.1 - 8, 9 1039 9051797
BC143295.1 - 9, 10 1116
BC143299.1 - 8, 9 1039 9051803
BC143330.1 - 7, 8 870 9051835
BC143333.1 - 8, 9 959
BC143334.1 - 7, 8 952
BX537539.1 - 1, 2 97
EU295940.1 - 6, 7 776

View Full Product Details